Copper(II) sulfate pentahydrate ≥98.0%, granular FCC
Supplier: Spectrum Chemical Mfg. Corp.
|
|
Copper Sulfate, Pentahydrate, Granular, FCC is used as a fungicide, algaecide and feed supplement. Find a high quality inventory of Copper Copper Sulfate, Pentahydrate, Granular, FCC. Approved by the Food Chemical Codex, it is used as a fungicide, algaecide, and feed supplement, and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Skull and crossbones;Exclamation mark
UN3077
Formula:
CuSO₄·5H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00149681 CAS Number: 7758-99-8 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...