Silicon dioxide ≥99.0%, colloidal Ph. Eur., NF
Supplier: Spectrum Chemical Mfg. Corp.
S1510-100GM
S1510-500GM
S1510-5KGBL
75812-336EA
323.68
CAD
75812-336
75812-338
75812-340
Silicon dioxide ≥99.0%, colloidal Ph. Eur., NF
Silicium dioxide
Colloidal Silicon Dioxide, NF, EP is used both in the food and pharmaceutical industries as a powder flow helper in the production of tablets. All Spectrum Chemical NF, EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
SiO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 7631-86-9 |
Specification Test Results
Assay (Ignited Basis) | 99.0 - 100.5 % |
pH (Nf) | 3.5 - 5.5 |
pH (Ep) | 3.5 - 5.5 |
Loss On Drying | max. 2.5 % |
Loss On Ignition (Nf) | max. 2.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...