Cod liver oil USP

Supplier: SPECTRUM CHEMICAL MFG CORP CA

CO112-4LTGL CO112-4LTGL
CA95032-672CS 2699 CAD
CA95032-672 CA95032-674
Cod liver oil USP
Cod liver oil

Cod Liver Oil, USP is an oil from the liver of Cod fish. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Storage Temperature: Ambient
MDL Number: MFCD00146493
CAS Number: 8001-69-2

Order Now

Specification Test Results

Vitamin A Content 180 - 750 Ug/G
Vitamin D Content 1.5 - 6.25 Ug/G
Color to Pass Test
Specific Gravity @ 25°C 0.918-0.927
Acid Value To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR