Citric acid monohydrate ≥99.5%, granular FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid

C1283-500GM C1283-12KG C1283-50KGBL
95032-600EA 352.26 CAD
95032-600 75810-466 75810-468
Citric acid monohydrate ≥99.5%, granular FCC
Citric acid monohydrate

Citric Acid, Monohydrate, Granular, FCC is widely used as acidity regulator and presevative in food production and as an acidity regulator and antioxidant in beverages. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formula: C₆H₈O₇.H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149972
CAS Number: 5949-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (C₆H₈O₇ ;Anhyd.,Basis) 99.5-100.5 %
Lead (Pb) max.0.5 mg/kg
Oxalate (C₂O₄) To Pass Test
Readily Carbonizable Substances To Pass Test
Residue On Ignition max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR