Citric acid, anhydrous ≥99.5%, granular USP

Supplier: Spectrum Chemical Mfg. Corp.

CI133-25KGBL
75810-940EA 2502.72 CAD
75810-940
Citric acid, anhydrous ≥99.5%, granular USP
Citric acid

Anhydrous Citric Acid, Granular, USP belongs to a class of drugs known as urinary alkalinizers that are used to treat certain metabolic problems (acidosis) caused by kidney disease. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formula: HOC(COOH)(CH₂COOH)₂
Storage Temperature: Ambient
MDL Number: MFCD00011669
CAS Number: 77-92-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Anhydrous Basis) 99.5 - 100.5 %
Clarity of Solution To Pass Test
Color of Solution To Pass Test
Water max. 1.0 %
Residue On Ignition max. 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR