Citric acid, anhydrous ≥99.5%, granular FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid

C1281-12KG C1281-45KGBL
75810-786EA 1993.01 CAD
75810-786 75810-788
Citric acid, anhydrous ≥99.5%, granular FCC
Citric acid

Citric Acid, Anhydrous, Granular, FCC is the most widely used organic acid and pH-control agent in foods and beverages. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formula: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Boiling Pt: 310
Storage Temperature: Ambient
MDL Number: MFCD00011669
CAS Number: 77-92-9
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (C₆H₈O₇ ;Anhyd.,Basis) 99.5 - 100.5 %
Lead (Pb) max. 0.5 mg/kg
Oxalate (C₂O₄) To Pass Test
Readily Carbonizable Substances To Pass Test
Residue On Ignition max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR