Chondroitin sulfate sodium salt ≥90.0% USP dietary supplement

Supplier: Spectrum Chemical Mfg. Corp.

C1610-100GM C1610-2.5KG C1610-500GM
75810-842EA 529.49 CAD
75810-842 75810-844 75810-846
Chondroitin sulfate sodium salt ≥90.0% USP dietary supplement
Chondroitin sulfate sodium salt

Chondroitin Sulfate Sodium Salt, USP Dietary Supplement.


All Spectrum Chemical USP Dietary Supplement grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).

Storage Temperature: Ambient
MDL Number: MFCD00130792
CAS Number: 9082-07-9

Order Now

Specification Test Results

Chondroitin Sulfate Sodium (Dried Basis) 90.0 - 105.0 %
Specific Rotation, [A]D25 -20.0 to -30.0
Residue On Ignition 20.0 - 30.0 %
pH (1 in 100) 5.5 - 7.5
Clarity and Color of Solution To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR