Cesium chloride, BiotechGrade

Supplier: Spectrum Chemical Mfg. Corp.

C1252-100GM C1252-25GM
75810-776EA 666.29 CAD
75810-776 75810-778 75810-780 75810-782
Cesium chloride, BiotechGrade
Cesium chloride

Cesium Chloride, BiotechGrade is a colorless inorganic compound and a solid. It is used in isopynic centrifugation as well as in analytical chemistry as a reagent.

Formula: CsCl
Storage Temperature: Ambient
MDL Number: MFCD00010955
CAS Number: 7647-17-8

Order Now

Specification Test Results

Appearance Fine,White,Cryst
Assay 99.90 % min.
Absorbance of a 50 % (W/V) Soln.@ 260 NM max. 0.02
Insoluble Matter To Pass Test
Heavy Metals (As Pb) max. 0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR