Cedar leaf oil 60% FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
Methyl 2-hexynoate
Cedar Leaf Oil, FCC is considered generally safe by the FDA as a food additive preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Flame;Exclamation mark
UN1169
Storage Temperature:
Ambient |
MDL Number:
MFCD00146327 CAS Number: 8007-20-3 UN: 1169 ADR: 3,III |
Specification Test Results
Assay (Ketones As Thujone) | 60 % |
Angular Rotation | -7 to -14 |
Refractive Index @ 20°C | 1.456- 1.460 |
Solubility in Alcohol | To Pass Test |
Specific Gravity @ 25°C | 0.906- 0.916 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...