DL-Camphor (synthetic) USP
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
(±)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, Camphor, 1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one, DL-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (±)-1,7,7-Trimethyl-2-norbornanone
CA185-12KG
CA185-25KGBL
75810-922EA
3023.96
CAD
75810-922
75810-924
DL-Camphor (synthetic) USP
DL-Camphor
Camphor, Synthetic, USP is a tablet porosity modifier excipient.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₁₀H₁₆O Density: 0.992 Storage Temperature: Ambient |
MDL Number:
MFCD00074738 CAS Number: 76-22-2 UN: 2717 ADR: 4.1,III |
Specification Test Results
Melting Range | 174° - 179°C |
Specific Rotation | Optically Inacti |
Appearance of Solution | To Pass Test |
Nonvolatile Residue | max. 0.05% |
Halogens | max. 0.035% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...