Calcium stearate FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Stearic acid calcium salt
CALCIUM STEARATE, FCC, also known as E470, is used as a flow agent in powders. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
All Spectrum Chemical FCC grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formula:
C₃₆H₇₀CaO₄ MW: 607.03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00036390 CAS Number: 1592-23-0 |
Specification Test Results
Assay (Dried Basis) | 9.0 - 10.5 % |
Free Fatty Acids | max.3.0 % |
Lead (Pb) | max. 2 mg/kg |
Loss On Drying | max. 4.0 % |
Identification | To Pass Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...