L(+)-Calcium lactate pentahydrate ≥98.0% USP, Ph. Eur., FCC
Supplier: Spectrum Chemical Mfg. Corp.
C2106-12KG
C2106-2.5KG
C2106-500GM
C2106-50KGBL
75810-868EA
2365.5
CAD
75810-868
75810-870
75810-872
75810-874
L(+)-Calcium lactate pentahydrate ≥98.0% USP, Ph. Eur., FCC
L(+)-Calcium lactate pentahydrate
Calcium Lactate, Pentahydrate, USP, FCC, EP is a filler/diluent excipient.
All Spectrum Chemical USP, FCC, EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formula:
C₆H₁₀CaO₆·5H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150344 CAS Number: 5743-47-5 |
Specification Test Results
Assay (Dried Basis) | 98.0 - 101.0% |
Loss On Drying | 22.0 - 27.0% |
Volatile Fatty Acids | To Pass Test |
Heavy Metals | max. 10 ppm |
Lead (Pb) | max. 2 mg/kg |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...