Calcium hydroxide ≥95.0%, powder USP, Ph. Eur., BP, JP
Supplier: Spectrum Chemical Mfg. Corp.
|
|
CA150-12KG
CA150-2.5KG
CA150-500GM
CA150-45KGBL
700000-928EA
1691.07
CAD
700000-928
95032-288
95032-290
75810-904
Calcium hydroxide ≥95.0%, powder USP, Ph. Eur., BP, JP
Calcium hydroxide
Calcium Hydroxide, Powder, USP, EP, BP, JP is used to adjust pH of pharmaceuticals.
All Spectrum Chemical USP, EP, BP, JP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formula:
Ca(OH)₂ Storage Temperature: Ambient |
MDL Number:
MFCD00010901 CAS Number: 1305-62-0 UN: 3262 ADR: 8,III |
Specification Test Results
Assay | 95.0 - 100.5 % |
Acid-Insoluble Substances | max. 0.5 % |
Carbonate | To Pass Test |
Heavy Metals | max. 20 ppm |
Arsenic | max. 4 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...