Calcium hydroxide ≥95.0%, powder FCC

Supplier: Spectrum Chemical Mfg. Corp.

C1116-12KG C1116-45KGBL
75810-764EA 1670.85 CAD
75810-764 75810-766
Calcium hydroxide ≥95.0%, powder FCC
Calcium hydroxide

Calcium Hydroxide, Powder, FCC is used extensively in the food industry to clarify solutions, as a digestion aid, baking soda substitute and as a nutritional supplement because of its low toxicity. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Corrosion;Exclamation mark

Formula: Ca(OH)₂
Storage Temperature: Ambient
MDL Number: MFCD00010901
CAS Number: 1305-62-0
UN: 3262
ADR: 8,III

Order Now

Specification Test Results

Assay [Ca(OH)₂] 95.0 - 100.5 %
Acid-Insoluble Substances max. 0.5 %
Arsenic (As) max. 3 mg/kg
Carbonate To Pass Test
Fluoride max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR