Calcium-D gluconate, anhydrous ≥98.0%, powder USP

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: Calcium Gluconate, Calcium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, Gluconate Calcium, D-Gluconic acid calcium salt

C1939-500GM C1939-2.5KG
CA95032-272EA 391 CAD
CA95032-272 75810-862
Calcium-D gluconate, anhydrous ≥98.0%, powder USP
Calcium D-gluconate

Calcium Gluconate, Anhydrous, Powder, USP is used as a nutrient. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: HOH₂C(CH(OH))₄COOCa
MW: 430.38 g/mol
Melting Pt: 178
Storage Temperature: Ambient
MDL Number: MFCD00064209
CAS Number: 299-28-5
Merck Index: 13,01671

Order Now

Specification Test Results

Assay 98.0 - 102.0 %
Loss On Drying max. 3.0 %
Chloride (Cl) max. 0.07 %
Sulfate max. 0.05 %
Arsenic (As) max. 3 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR