Calcium citrate tetrahydrate ≥97.5%, powder FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: tri-Calcium dicitrate tetrahydrate, Calcium citrate tribasic tetrahydrate

CA142-12KG CA142-56.7KGBL
75810-900EA 3837.11 CAD
75810-900 75810-902
Calcium citrate tetrahydrate ≥97.5%, powder FCC
Calcium citrate tetrahydrate

Calcium Citrate, Tetrahydrate, Powder, FCC is an odorless white powder and the calcium salt of citric acid and is commonly used as a preservative and flavorant food additive. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₂H₁₀Ca₃O₁₄·4H₂O
MW: 570.5 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150786
CAS Number: 5785-44-4

Order Now

Specification Test Results

Assay [Ca₃(C₆H₅O₇)₂] 97.5 - 100.5 %
Loss On Drying 10.0 - 14.0 %
Fluoride max. 0.003 %
Lead (Pb) max. 2 mg/kg
Identification To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR