Calcium chloride dihydrate, BiotechGrade

Supplier: Spectrum Chemical Mfg. Corp.

C1086-12KGBL C1086-2.5KG C1086-500GM
75810-738EA 3333.33 CAD
75810-738 75810-740 75810-742
Calcium chloride dihydrate, BiotechGrade
Calcium chloride dihydrate

Calcium Chloride, Dihydrate, BiotechGrade is a white deliquescent compound which is commonly used as a reagent in biochemistry. Biological processes, including signal transduction, muscle contraction, maintenance of cell membranes and cell wall stability rely on calcium ions. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.


Spectrum offers highly pure reagents suitable for biochemical research and analysis.

Formula: CaCl₂·2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8

Order Now

Specification Test Results

Appearance White Crystals
Odor Odorless
Assay (CaCl₂[-2H₂O) 99.0 - 103.0 %
pH of a 5% Solution @ 25°C 4.5 - 9.2
Acidity / Alkalinity To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR