Calcium chloride, anhydrous ≥93.0%, pellets FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

C1977-12KG C1977-45KGBL
75810-864EA 1249.94 CAD
75810-864 75810-866
Calcium chloride, anhydrous ≥93.0%, pellets FCC
Calcium chloride

Calcium Chloride, Anhydrous, Pellets, FCC is used as a sequestrant and firming agent in food products. It is considered as generally recognized as safe (GRAS) by the U.S. Food and Drug Administration. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Exclamation mark

Formula: CaCl₂
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4

Order Now

Specification Test Results

Assay 93.0 - 100.5 %
Acid-Insoluble Substances max. 0.02 %
Arsenic (As) max. 3 mg/kg
Fluoride max. 0.004 %
Lead (Pb) max. 5 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR