Calcium carbonate 98,0, powder_light USP
Supplier: SPECTRUM CHEMICAL MFG CORP CA
|
|
Synonyms:
Marble
CA130-12KG
CA130-45KGBL
CA130-25KGBL
CA700001-806EA
3538.99
CAD
CA700001-806
CA700001-808
75810-896
Calcium carbonate 98,0, powder_light USP
Calcium carbonate
Calcium Carbonate, Light Powder, USP is a buffering agent excipient.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
CaCO₃ MW: 100.09 g/mol Melting Pt: 825 Storage Temperature: Ambient |
MDL Number:
MFCD00010906 CAS Number: 471-34-1 Merck Index: 13,01658 |
Specification Test Results
Assay (Dried Basis) | 98.0 - 100.5 % |
Loss On Drying | max. 2.0 % |
Acid-Insoluble Substances | max. 0.2 % |
Fluoride | max. 0.005 % |
Arsenic (As) | max. 3 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...