D-(+)-Biotin ≥97.5%, powder FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7

BI115-100GMBL BI115-1GM BI115-25GM BI115-500MG BI115-5GM
75810-708EA 5831.39 CAD
75810-708 75810-710 75810-712 75810-714 75810-716
D-(+)-Biotin ≥97.5%, powder FCC
D(+)-Biotin

Biotin, Powder, FCC is often recommended as a dietary supplement for strenghening hair and nails. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: C₁₀H₁₆N₂O₃S
Storage Temperature: Refrigerator
MDL Number: MFCD00005541
CAS Number: 58-85-5

Order Now

Specification Test Results

Assay (C₁₀H₁₆N₂O₃S) 97.5 - 100.5 %
Melting Range 229 - 232°C
Optical Rotation +89 to +93
Lead (Pb) max. 2 mg/kg
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR