D-(+)-Biotin ≥97.5%, powder USP

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: Vitamin HVitamin B85-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acidD-BiotinVitamin B7

B1103-500MG B1103-1GM B1103-1GM B1103-25GM B1103-5GM B1103-500GMBL B1103100GMBL B1103-25GM B1103-1KGBL B1103-500MG B1103-5GM
CA89085-278EA 9849.93 CAD
700009-418 CA89085-278 CA89085-276 89085-276 CA89085-274 89085-278 CA89085-272 89085-272 89085-274 89085-280 75810-670
D-(+)-Biotin ≥97.5%, powder USP
D(+)-Biotin

Biotin, Powder, USP is a water-soluble B vitamin. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₀H₁₆N₂O₃S
MW: 244.31 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00005541
CAS Number: 58-85-5

Order Now

Specification Test Results

Assay 97.5 - 102.0 %
Specific Rotation, [A]D25 +89° to +93°
Related Compounds To Pass Test
Identification To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR