Benzoic acid ≥99.5% FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
B1102-12KG
B1102-45KGBL
75810-666EA
2647.29
CAD
75810-666
75810-668
Benzoic acid ≥99.5% FCC
Benzoic acid
Benzoic Acid, FCC is used as a food preservative inhibiting the growth of mold, yeast and some bacteria. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
GHS hazard : Exclamation mark
Formula:
C₇H₆O₂ MW: 122.12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002398 CAS Number: 65-85-0 |
Specification Test Results
Assay (C₇H₆O₂) | 99.5 - 100.5 % |
Solidification Point | 121° - 123°C |
Lead | max. 2.0 mg/kg |
Readily Carbonizable Substances | To Pass Test |
Readily Oxidizable Substances | To Pass Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...