L(+)-Ascorbic acid ≥99.0%, crystalline powder USP
Supplier: SPECTRUM CHEMICAL MFG CORP CA
Synonyms:
Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
Spectrum-labeled kosher products with the Kosher Supervision of America symbol are a firm guarantee to all Spectrum customers that the products bearing the symbol are in full compliance with the most demanding of Kosher and Pareve standards.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₆H₈O₆ MW: 176.13 g/mol Boiling Pt: 553 Storage Temperature: Ambient |
MDL Number:
MFCD00064328 CAS Number: 50-81-7 Merck Index: 13,00837 |
|
Specification Test Results
Assay | 99.0 - 100.5% |
Specific Rotation [A]D | +20.5° to +21.5 |
Residue On Ignition | max. 0.1% |
Heavy Metals (As Pb) | max. 0.002% |
Identification | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...