L(+)-Ascorbic acid ≥99.0%, crystalline powder FCC

Supplier: Spectrum Chemical Mfg. Corp.

A1371-100GM A1371-12KG A1371-50KGBL
75810-528EA 362.37 CAD
75810-528 75810-530 75810-532
L(+)-Ascorbic acid ≥99.0%, crystalline powder FCC
L(+)-Ascorbic acid

Ascorbic Acid, Crystalline Powder, FCC is commonly used as antioxidant food additive. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₆H₈O₆
Storage Temperature: Ambient
MDL Number: MFCD00064328
CAS Number: 50-81-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c03499908e269560d680b0d7c4651f97a367aa0&vid=73a720d6900e713a98164022273a8afa6114ad7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (C₆H₈O₆) 99.0 - 100.5 %
Specific Rotation (C=10; Water)) +20.5 to +21.5
Lead (Pb) max. 2 mg/kg
Residue On Ignition max. 0.1 %
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR