Anise oil FCC

Supplier: Spectrum Chemical Mfg. Corp.

AN119-51002 AN119-2.5LT
CA11024-276EA 1271.99 CAD
CA11024-276 75810-642
Anise oil FCC
Anise oil

Anise Oil, FCC is used as a flavoring agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Density: 0.980
Storage Temperature: Ambient
MDL Number: MFCD00130517
CAS Number: 8007-70-3

Order Now

Specification Test Results

Angular Rotation @25°C -2 to +1
Refractive Index @ 20°C 1.553 - 1.560
Solidification Point 15 °C min.
Solubility in 90% Alcohol To Pass Test
Specific Gravity @ 25°C 0.978- 0.988


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR