Diammonium hydrogen orthophosphate ≥96.0% FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate
Ammonium Phosphate Dibasic, FCC is a food ingredient used as an acidity regulator and raising agent. It is listed on the FDA's Generally Recognized As Safe (GRAS) list of substances. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Formula:
(NH₄)₂HPO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00010891 CAS Number: 7783-28-0 |
Specification Test Results
Assay [(NH₄)₂HPO₄] | 96.0 - 102.0 % |
Arsenic (As) | max. 3 mg/kg |
Fluoride | max. 10 mg/kg |
Lead (Pb) | max. 4 mg/kg |
Identification | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...