Ammonium persulfate (APS), crystals, BiotechGrade

Supplier: Spectrum Chemical Mfg. Corp.

A1227-100GM A1227-12KGBL A1227-2.5KG A1227-25GM A1227-500GM
75810-456EA 199.19 CAD
75810-456 75810-458 75810-460 75810-462 75810-504
Ammonium persulfate (APS), crystals, BiotechGrade
Ammonium peroxodisulphate (APS)

Ammonium Peroxydisulfate, Crystal, BiotechGrade this highly water soluble oxidizing agent is a catalyst for acrylamide gel polymerization when used in conjunction with Tetramethylethylenediamine. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.


GHS hazard : Flame over circle;Exclamation mark;Health hazard
UN1444

Formula: (NH₄)₂S₂O₈
Storage Temperature: Ambient
MDL Number: MFCD00003390
CAS Number: 7727-54-0
UN: 1444
ADR: 5.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82a373f21d34e63dc94c0598f3f33ba6bbfdb2b&vid=2b6d196dbf07ce4dd528ac7b4f8456678accd3ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Whi
Assay 98.0 %
Insoluble Matter max. 0.005 %
Residue After Ignition max. 0.05 %
Titrable Free Acid max. 0.04 Meq


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR