Ammonium chloride ≥99.0%, granular FCC

Supplier: Spectrum Chemical Mfg. Corp.

A1167-12KG A1167-45KGBL
75810-452EA 2218.72 CAD
75810-452 75810-454
Ammonium chloride ≥99.0%, granular FCC
Ammonium chloride

Ammonium Chloride, Granular, FCC is an ingredient in nutritive media for yeasts and many microorganisms. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Exclamation mark

Formula: NH₄Cl
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Dried Basis) 99.0 % min.
Lead (Pb) max.4 mg/kg
Loss On Drying max. 0.5 %
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR