Ammonia solution 27 - 30% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

A1293-200LTBL A1293-20LT A1293-500MLGL
75810-516EA 3815.7 CAD
75810-516 75810-518 75810-520
Ammonia solution 27 - 30% FCC
Ammonia solution

Ammonia Solution, FCC is a solution of Ammonia in water. It has many uses, but in food production it is used primarily as a leavening agent. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Corrosion;Exclamation mark
UN2672

Formula: NH₃
Density: 0.900
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Assay (NH₃, By Wt.) 27.0 - 30.0 %
Lead (Pb) max. 0.5 mg/kg
Nonvolatile Residue max. 0.02 %
Readily Oxidizable Substances To Pass Test
Identification To Pass Test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR