Aluminium potassium sulfate dodecahydrate 99.5-100.5%, crystals FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Aluminium Potassium sulphate, 12-Hydrate, Aluminium Potassium sulfate, 12-Hydrate, Aluminum potassium sulfate dodecahydrate, Aluminum potassium sulphate dodecahydrate, Aluminium potassium bis(sulphate) dodecahydrate, Potassium alum, Alum potassium, Aluminium potassium bis(sulfate) dodecahydrate

A1107-500GM A1107-12KG A1107-50KGBL
95031-744EA 308.02 CAD
95031-744 75810-444 75810-446
Aluminium potassium sulfate dodecahydrate 99.5-100.5%, crystals FCC
Aluminium potassium sulphate dodecahydrate

Aluminum Potassium Sulfate, Crystal, FCC is commonly used as an additive to baking powder to provide a second leavening phase at high temperatures. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: AlK(SO₄)₂·12H₂O
MW: 474.39 g/mol
Melting Pt: 92.5 °C/198.5 °F
Density: 1.757
Storage Temperature: Ambient
MDL Number: MFCD00149143
CAS Number: 7784-24-9
Merck Index: 12,00373

Order Now

Specification Test Results

Assay [Alk(SO₄)₂[-12H₂O] 99.5-100.5 %
Ammonium Salts To Pass Test
Fluoride max. 0.003 %
Lead (Pb) max. 3 mg/kg
Selenium (Se) max. 0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR