(±)-α-Lipoic acid ≥99.0% USP dietary supplement

Supplier: Spectrum Chemical Mfg. Corp.

L1506-100GM L1506-1KGBL L1506-25GM L1506-5GM
75811-500EA 3346.91 CAD
75811-500 75811-502 75811-504 75811-506
(±)-α-Lipoic acid ≥99.0% USP dietary supplement
(±)-α-Lipoic acid

Alpha Lipoic Acid, USP Dietary Supplement.


GHS hazard : Exclamation mark

Formula: C₈H₁₄O₂S₂
Storage Temperature: Refrigerator
MDL Number: MFCD00005474
CAS Number: 1077-28-7

Order Now

Specification Test Results

Assay (Dried Basis) 99.0 - 101.0%
Melting Range 60.0° - 62.0°C
Specific Rotation - 1.0° to +1.0
6,8-Epitrithiooctanoic Acid max. 0.1%
Heavy Metals max. 0.001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR