Adipic acid ≥99.6% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Hexanedioc acid, 1,6-Hexanedioic acid

AD130-100GM AD130-12KG
75810-606EA 161.03 CAD
75810-606 75810-608
Adipic acid ≥99.6% FCC
Adipic acid

Adipic Acid, FCC is used as a food ingredient as a flavorant and gelling aid and as an acidulant in baking powders. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formula: HOOC(CH₂)₄COOH
Storage Temperature: Ambient
MDL Number: MFCD00004420
CAS Number: 124-04-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58ac39e8ebd69527d91cb2d15a469a2a69506785&vid=ef907d9f9fd34903274c42b99d1d1ac1a8db6dce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (C₆H₁₀O₄) 99.6 - 101.0 %
Melting Range 151.5 - 154 °C
Water max. 0.2 %
Lead (Pb) max. 2 mg/kg
Residue On Ignition max. 0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR