Acetone ≥99.5% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2-PropanoneDMKPropanoneDimethyl ketone

A1041-200LTBL A1041-4LTGL-CS A1041-100ML A1041-4LTPL A1041-4LTGL A1041-1LT A1041-500MLGL A1041-4LTPL-CS A1041-500ML-CS A1041-20LT A1041-1LT-CS6
75810-428EA 2654.24 CAD
75810-428 75810-426 75810-438 75810-424 75810-436 75810-422 75810-434 75810-442 75810-432 75810-440 75810-430
Acetone ≥99.5% FCC
Acetone

Acetone, FCC also known as 2-propanone, is an organic compound and a colorless, flammable liquid. Due to its miscibility with water, it serves as an important solvent. The U.S. Food and Drug Administration (FDA) has determined acetone is safe for use as an indirect food additive in adhesives and food-contact coatings and is regarded as a Generally Recognized as Safe (GRAS) substance at certain concentrations. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Flame;Exclamation mark;Health hazard
UN1090

Formula: CH₃COCH₃
MW: 58.08 g/mol
Density: 0.788
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
UN: 1090
ADR: 3,II
https://ca.vwr.com/stibo/web/std.lang.all/06/07/720607.jpg

Order Now

Specification Test Results

Assay 99.5 - 100.5 %
Distillation Range Within 1 °C
Refractive Index 1.358 - 1.360
Solubility in Water To Pass Test
Acidity (As CH₃COOH) max. 0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR