Acetic acid glacial FCC

Supplier: Spectrum Chemical Mfg. Corp.

A1009-200LTBL A1009-20LTPL
75810-418EA 5895.06 CAD
75810-418 75810-420
Acetic acid glacial FCC
Acetic acid

Acetic Acid, Glacial, FCC is used in the pickling of vegetables and other foods. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Flame;Exclamation mark;Corrosion
UN2789

Formula: H₃CCOOH
Density: 1.049
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
UN: 2789
ADR: 8.3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5 - 100.5 %
Solidification Point 15.6°C min.
Nonvolatile Residue max. 0.005 %
Lead (Pb) max. 0.5 mg/kg
Readily Oxidizable Substances To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR