Acesulfame potassium ≥99.0% NF
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
6-Methyl-1,2,3-oxathiazin-4(3H)-one 2,2-Dioxide Potassium Salt, Potassium 6-methyl-4-oxo-4H-1,2,3-oxathiazin-3-ide 2,2-dioxide, Acesulphame K, Potassium 6-Methyl-1,2,3-oxathiazin-4(3H)-one 2,2-Dioxide, Acesulfame K
A3974-100GM
A3974-1KGBL
A3974-25GM
A3974-500GM
75810-580EA
748.65
CAD
75810-580
75810-582
75810-584
75810-586
Acesulfame potassium ≥99.0% NF
Acesulfame potassium
Acesulfame Potassium, NF is a white crystallized powder.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formula:
C₄H₄KNO₄S MW: 201.24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00043833 CAS Number: 55589-62-3 |
Specification Test Results
Assay | 99.0 - 101.0 % |
Acidity or Alkalinity | To Pass Test |
Chromatographic Purity | To Pass Test |
Heavy Metals | max. 10 ppm |
Fluoride | max. 3 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...