Gum arabic, spray-dried powder NF

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Acacia, Acacia gum

GU115-2.5KG GU115-10 GU115-100GM
95031-586EA 1041.3 CAD
95031-586 CA11024-344 75811-296
Gum arabic, spray-dried powder NF
Gum arabic

Acacia, Spray-Dried Powder, NF is an excipient binder. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Storage Temperature: Ambient
MDL Number: MFCD00081264
CAS Number: 9000-01-5

Order Now

Specification Test Results

Solubility and Reaction To Pass Test
Botanical Characteristics To Pass Test
Microbial Limits To Pass Test
Water Determination max. 15.0 %
Total Ash max. 4.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR