Flow Meter, Nitrogen, with Metering Valve, Chemglass
Supplier: Chemglass
CG-1605-A-10 CG-1605-A-12 CG-1605-A-14
16002-788EA 650.31 CAD
16002-788 16002-790 89028-648
Flow Meter, Nitrogen, with Metering Valve, Chemglass
Flow Meters
Precision machined clear acrylic Nitrogen flowmeter with brass metering valve and Buna-N seals for use with Chemglass process reactors.
  • Front of flowmeter has a blue scale indicating the L/min. of air
  • HDPE hose connection is for use with 5/16” ID tubing
Delivery information: Unit is supplied complete with a 1/2" O.D. mounting arm for ease in clamping to support frame. Flowmeters with larger L/min. ranges are available.