Hydroxy naphthol blue disodium salt ACS for calcium determination

Supplier: CHEM-IMPEX INTERNATIONAL, INC

Synonyms: Hydroxynaphthol blue sodium salt, Hydroxy napthol blue, Sodium (E)-3-hydroxy-4-((2-hydroxy-6-sulfo-3-sulfonatonaphthalen-1-yl)diazenyl)naphthalene-1-sulfonate, Sodium (E)-3-hydroxy-4-((2-hydroxy-6-sulpho-3-sulphonatonaphthalen-1-yl)diazenyl)naphthalene-1-sulphonate

22810-100G 22810-250G 22810-25G
CH22810-100GEA 150.1 CAD
CH22810-100G CH22810-250G CH22810-25G
Hydroxy naphthol blue disodium salt ACS for calcium determination
Hydroxy naphthol blue disodium salt

Used in the determination of calcium.


Synonyms: Hydroxy napthol blue disodium salt

Formula: C₂₀H₁₂N₂O₁₁S₃·2Na
MW: 598.479 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004075
CAS Number: 165660-27-5

Order Now

Specification Test Results

Appearance Dark blue crystals
Suitability for Calcium Determination To pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR