Peptide Vessels, Jacketed with Lower Drain Valves, Chemglass
Supplier: Chemglass
CG-1868-01 CG-1868-02 CG-1868-03 CG-1868-04
80071-402EA 3100.96 CAD
80071-402 80071-404 80071-406 80071-408
Peptide Vessels, Jacketed with Lower Drain Valves, Chemglass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Cylindrical, jacketed, peptide synthesis reaction vessel with coarse porosity fritted resin support.
  • The vessel is constructed with a flange having a groove formed in the sealing surface of the flange
  • Flange is also ground to permit a greased seal with any of the corresponding lids
  • Lower outlet is a CG-511 bottom drain valve
Cooling jacket is equipped with two removable hose connections to reduce the risk of injury while attaching or detaching flexible tubing.
Ordering information: Supplied complete with two polypropylene hose connections and twoMore Product Information