VWR® DURAC Plus Dry Block/Incubator Liquid-In-Glass Thermometer; Organic Liquid Fill
Supplier: VWR
VWR®
607010011 60701-0011
CA89095-780CS 3577.44 CAD
CA89095-780 89095-780
VWR® DURAC Plus Dry Block/Incubator Liquid-In-Glass Thermometer; Organic Liquid Fill
Thermometers Analog Thermometers
Ideal for use in all standard heating blocks and water baths, these thermometers contain blue organic liquid fill against lead-free white back glass.
  • Noncalibrated
  • Traceable to NIST
  • Include multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of accuracy and traceability
  • Tested and inspected by our USA quality team
Ordering information: This thermometer is noncalibrated. If an individual Thermometer Calibration Report is required, factory calibrated thermometers or custom calibration services are available. For a calibrated thermometer, contact your Avantor representative for more information.