Louvered Hanging Systems, Akro-Mils
Supplier: Akro-Mils
30118 30136 30161 30661 30676 98600 98636 30118BEIGE 30118TEXWHT 30618 30636 30651 30653
75854-430EA 445.53 CAD
75854-430 75854-432 75854-436 75854-446 75854-448 75854-596 75854-598 75854-772 75854-774 75855-888 75855-890 75855-896 75855-900
Louvered Hanging Systems, Akro-Mils
Furniture Shelves
Louvered Panels, Floor Racks, and Bench Racks allow bins to securely hang, keeping parts and supplies organized and out of the way.
  • Formed from 16-gauge cold-rolled steel for strength and durability
  • Steel rails securely support an assortment of Akro-Mils bins and containers (sold separately)
  • Durable powder-coated finish stands up to heavy-duty service
  • Made in the USA
Wall-mount louvered panels keep parts up off the ground and free up floor space
All ReadySpace® panels have a clean, sleek look with a durable white, powder-coated finish. These systems are designed for use in hospitals, labs, and medical offices. They are also available in wall, floor, or benchtop formats.
Floor racks can be placed near work areas to improve productivity. Bench racks keep vital tools and supplies at eye level and within reach.