Screw Compressor Clamp, Humboldt
Supplier: Humboldt Manufacturing
H-8680 H-8720 H-8730 H-8700
21715-016EA 32.55 CAD
21715-016 89370-250 89370-266 89370-270
Screw Compressor Clamp, Humboldt
Clamps Tubing and Sanitary Clamps
Screw Compressor Clamps are used to control the flow in flexible tubing.
  • Clamps have a machined adjustment screw to permit accurate regulation of the flow
  • Rounded corners help prevent cutting of the tubing
  • Clamps feature brass construction, plated for corrosion resistance and long life