National™ Centrifugal Filters, Non sterile, 2 ml, Thermo Scientific
Supplier: Thermo Scientific
F2520-3 F2520-5 F2520-7 F2520-8 F2520-1 F2520-2 F2520-4 F2520-6
66064-484PK 386.22 CAD
66064-484 66064-472 66064-474 66064-476 66064-478 66064-480 66064-482 66064-470
National™ Centrifugal Filters, Non sterile, 2 ml, Thermo Scientific
Filters Centrifugal Filters
Filter volumes up to 2 ml with these National™ centrifugal filters.
  • 5000×g maximum centrifugal force
  • Non sterile
  • Bulk packed
Filters feature a virgin polypropylene filter housing with capped receiver tube, and can be used with any benchtop or floor model laboratory centrifuge.