Straight Wall Container, Akro-Mils
Supplier: Akro-Mils
37208GREY 37278GREY 37288GREY 37608GREY 37612GREY 37616GREY 37672GREY 37676GREY 37678GREY 37682GREY 37686GREY 37688GREY
75855-594CS 1090.12 CAD
75855-594 75855-598 75855-602 75855-606 75855-610 75855-614 75855-618 75855-622 75855-626 75855-630 75855-634 75855-638
Straight Wall Container, Akro-Mils
Boxes Storage Boxes
Heavy-duty Straight Wall Containers let users maximize space in storage or while shipping
  • Uniform 1" wall thickness for secure stacking
  • Straight walls provide efficient cubing in trucks and on pallets
  • Made in the USA
Available as all solid, all mesh, or solid bottom-mesh sides. Solid-sided containers have smooth inner walls for easy, sanitary cleaning. Mesh-sided containers have an open 0.72x0.9" mesh pattern which allows high visibility and good airflow. Open handles on all four sides. Strong 1” wide stacking rims and external ribbing provide added strength.
Straight walls provide efficient cubing in trucks and on pallets.