Dissolution Vessels and Centering System Accessories, Quality Lab Accessories
Supplier: Quality Lab Accessories
Agilent Caliper/Zymark QLA QLA Innovative Product Hanson Sotax Distek Erweka CALEVA
GLA500-AD GLA9PK-01 GLA9CR-DK GLA900-CA GLA9PK-AUNP GLA9PK-AH GLA02K-HR GLA900-AST GLA900-DKAP PLA900-AV GLA900-AEW GLA500-AV GLA900-UNP GLA9CR-AZM GLA900-AC GLA900-01 GLA900-PT ASTM9PK-AUNP GLA02K-01 GLA9CR-AD ASTM900-VK ASTM900-AUNP PLA900-01 ASTM900-UN ASTM900-UNP PLA900-EW GLA900-AUNP GLA02K-EW GLA900-AH GLA500-01 VESSTD-08H GLA9PK-DK GLA500-HR ASTM900-AVK GLA9CR-ZM GLA900-AV GLA900-ST GLA9PK-AV GLA500-DK PLA900-AD GLA9PK-HR GLA900-AD GLA9PK-VK ASTM9PK-UNP PLA900-HR GLA02K-AD PLA900-AP GLA900-HR GLA900-APT PLA900-AH ASTM900-UNAM GLA9PK-UNP GLA900-DKP GLA900-VK VESSTD-06H GLA900-DK GLA900-EW GLA02K-DK PLA900-PT PLA900-DK
11010-410EA 449.38 CAD
11010-410 75849-210 11010-332 11010-412 11010-334 89080-332 75849-214 89080-330 93000-978 75849-058 75849-212 89049-302 93000-976 75849-218 75849-216 75849-138 11010-414 11010-336 11010-338 93000-982 75849-140 75849-060 11010-366 11010-368 11010-362 11010-364 11010-406 11010-408 11010-404 75849-208 93000-974 89039-830 89173-100 11010-354 11010-356 89039-836 11010-350 11010-352 11010-314 11010-358 11010-316 89132-946 89132-948 89039-842 89080-324 11010-344 89133-036 11010-346 75849-142 75849-148 11010-340 75849-224 89133-076 11010-342 89133-074 75849-228 75849-226 89173-122 89080-328 75849-150
Dissolution Vessels and Centering System Accessories, Quality Lab Accessories
Dissolution Apparatus
QLA manufactures a wide selection of Precision Molded and Standard Glass Vessels for most dissolution testers.
  • All glass vessels are made of borosilicate glass in the USA
  • Vessels are available in clear and amber
  • UltraCenter Precision, Standard and Apex Vessels available with both glass and plastic rims
Good dissolution vessels are critical in obtaining correct and consistent results
UltraCenter Precision Vessels are manufactured using a unique molded process which provides a precise and consistent inside diameter and spherical radius resulting in more reproducible dissolution test results. They are compatible with most dissolution baths. QLA's molded Apex vessels are patent pending.
We have the solution if you are having problems with vessel centering. The QLA AccuCenter Bracket provides ultimate centering accuracy on most dissolution baths (both old and new). It is adjustable and designed to center the vessel from the inside diameter and not the outside. When used with our UltaCenter Precision Vessels with glass or plastic rims, extreme centering accuracy can be achieved. Note: Standard glass vessels cannot be used in the AccuCenter Bracket.
Plastic Rimmed Vessels were designed to help minimize glass rim breakage that can occur in the lab. All QLA plastic rim vessels are dishwasher safe.
All vessels meet the physical dimensions of the USP and have a hemispherical bottom which should be perfectly rounded with no obvious defects. QLA glass vessels are acid etched, or laser marked on plastic rim, with a unique serial number and come with a Certificate of Conformance (COC), where applicable.
Certifications: All vessels are supplied with a Certificate of Conformance (COC) stating that they are USP compliant, where applicable, and that they meet machine manufacturer specifications. A Certificate of Analysis (COA) is available at an extra cost.