KIMBLE® HI-VAC® T-Type Valve with PTFE Plug, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
826800-0004 826800-0008
KT826800-0004EA 142.22 CAD
KT826800-0004 KT826800-0008
KIMBLE® HI-VAC® T-Type Valve with PTFE Plug, Kimble Chase, DWK Life Sciences
Valves Vacuum Valves
T-valve configuration, without tip o-ring exposure, allows closure of the bottom arm while vacuum or gases continue to flow through the sidearms.
  • Designed with a precision external thread for fine control
  • Valve can be used as a bleed up to 1/2 turn
  • Full open is at 11/2 turns
The shaft is non-rotating and the primary seal is made with a FKM o-ring on a tapered seat. PTFE shaft can be used continuously at system temperatures of 230°C (446°F) or intermittently at 275°C (527°F). Manufactured from 33 expansion, low extractable borosilicate glass conformingMore Product Information