VWR® Laboratory Bottles, Square, Wide Neck, with GL80 Thread
Supplier: VWR International
75871-304 75871-306 75871-308 75871-310 75871-420 75871-422 75871-424 75871-426
75871-304CS 340.75 CAD
75871-304 75871-306 75871-308 75871-310 75871-420 75871-422 75871-424 75871-426
VWR® Laboratory Bottles, Square, Wide Neck, with GL80 Thread
Bottles Glass Bottles
Made from borosilicate 33 expansion glass, these bottles are available in both Clear and Amber color formats.
  • Excellent chemical resistance, high temperature resistance
  • Extra wide opening allows easy cleaning
  • Minimal thermal expansion provides high resistance to temperature changes
  • Polypropylene GL 80 caps included
The large opening enables easy storage and transfer of substances. Wide mouth openings also accommodate large volume funnels and gives easy access to scoops or spatulas. Amber bottles help protect light sensitive substances.