Molybdate 3 Reagent Solution, Hach
Supplier: Hach
199532
199549
199526
CA1995-32EA
73.61
CAD
CA1995-32
CA1995-49
CA11016-694
Molybdate 3 Reagent Solution, Hach
Environmental Test Kits
For low and ultra-low range silica determination by the Heteropoly Blue method
Hach methods 8282 and 8186.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.