Cogent Bidentate C18™ HPLC Columns, MicroSolv Technology Corporation
Supplier: MicroSolv Technology Corp
4001805P2 4001810P2 4001815P2 40018-05P-1 4001825P 4001805P 4001815P 4001815P1 4001825P1 4001875P 4001805P3 4001810P4 4001815P3 4001875P2 40018-10P-3 40218-02P-2 40218-03P-2 40018-MP100 40018-MP150 40018-MP250 40018-MP50 40018-MP75 40018-P21-100 40018-P21-150 40018-P21-250 40018-P21-50 40018-P21-75 40018-SP100 40018-SP150 40018-SP250 40018-SP50 40018-SP75 40018-P30-150 40018-P30-250 40018-P30-50 40018-03P-3 40018-25P-3 40018-75P-1 40018-75P-3 40218-02P-3 40018-P30-75 40018-02P 40018-02P-2 40018-02P-3 40018-03P 40018-03P-1 40018-03P-2 40218-03P-1 40218-15P-1
97034-966EA 1661.23 CAD
97034-966 97034-972 97034-978 97055-188 97034-942 97034-964 97034-974 97034-976 97034-980 97034-982 97035-088 97035-090 97035-092 97035-094 97035-096 89220-016 10022-530 10022-532 10022-534 10022-536 10022-538 10022-540 10799-312 10799-314 75801-138 75801-140 75801-142 75801-144 75801-146 75801-148 75801-150 75801-152 75801-154 75801-156 75801-158 75801-160 75801-162 75801-164 75801-166 75839-512 75839-514 75839-516 75839-542 75839-586 75839-588 75839-590 75839-670 75839-696 75839-834 75839-838 75839-840 75839-842 75839-844 75839-846 75839-880 75839-882 75856-530 75856-532 75856-558
Cogent Bidentate C18™ HPLC Columns, MicroSolv Technology Corporation
Chromatography Columns
Using our proprietary bonding technology, the straight chain C18 hydrocarbon is bonded directly to the silica hydride surface with two separate points of attachment via silicon-carbon bonds.
  • Bond is virtually indestructable producing
  • stable, efficient and unique HPLC columns
  • Columns operate in RP, ONP, and ANP
  • Easy gradient analysis
This column is often used as a lab workhorse since it is so stable and the column lifetime is vastly greater than other C18 columns.
The virtually indestructible bond produces the most stable, efficient and unique HPLC columns on the market that operate in Reversed Phase (RP), Normal Phase (ONP) and Aqueous Normal Phase (ANP).
Less water is required for good retention and gradient analysis is so much easier. No end capping is used so this column can be used at very low pH with no damage to it and it is also much more stable when used with PIC or paired ion reagents in RP.