Ward's Science Essentials® Gray Gneiss
Supplier: Avantor
470226-218EA 6.15 CAD
470226-218 470226-312 470226-432
Ward's Science Essentials® Gray Gneiss
Specimens Rock Specimens Metamorphic Rocks
High grade metamorphosed rock showing bands of quartz, feldspar, and biotite.
  • Perfect sized specimens for hands on investigations
  • Multiple Pack Offerings to Suit Your Needs
Ward’s Science Essentials
Engage and inspire students to explore in-demand STEM careers with Ward’s Science Essentials. These multi-disciplinary activities kits allow you to easily build STEM concepts into every lesson – with plenty of a-ha moments along the way.
Experience
Create real-world scenarios in the classroom, helping students see the role of STEM in everyday life.